2-Bromophenyl methyl sulfone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromophenyl Methyl sulphone, 1-Bromo-2-(methylsulphonyl)benzene

19251-25G 19251-5G
76884-238EA 212.18 USD
76884-238 76884-240
2-Bromophenyl methyl sulfone ≥97%
2-Bromophenyl methyl sulphone

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368924
CAS Number: 33951-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR