(2-Chloro-5-isopropylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

19119-1G 19119-25G 19119-500MG 19119-5G
76884-216EA 142.17 USD
76884-216 76884-218 76884-220 76884-222
(2-Chloro-5-isopropylphenyl)boronic acid ≥97%
(2-Chloro-5-isopropylphenyl)boronic acid
Formula: C₉H₁₂BClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16295259
CAS Number: 875550-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR