2-(Methoxycarbonyl)-5-(trifluoromethoxy)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

19058-1G 19058-25G 19058-500MG 19058-5G
76884-200EA 263.11 USD
76884-200 76884-202 76884-204 76884-206
2-(Methoxycarbonyl)-5-(trifluoromethoxy)phenylboronic acid ≥95% research grade
2-(Methoxycarbonyl)-5-(trifluoromethoxy)phenylboronic acid
Formula: C₉H₈BF₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD12025990
CAS Number: 1217500-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR