1-Bromo-4-fluoro-2-methoxymethoxy-benzene 95%

Supplier: AOBChem USA

Warning

19033-1G 19033-25G 19033-5G 19033-500MG
76884-186EA 83.82 USD
76884-186 76884-188 76884-190 77645-070
1-Bromo-4-fluoro-2-methoxymethoxy-benzene 95%
1-Bromo-4-fluoro-2-methoxymethoxy-benzene
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20441852
CAS Number: 162269-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR