2,4-Difluoro-1-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2',4'-Difluorophenoxy)propane

18971-1G 18971-25G 18971-5G
76884-156EA 62.6 USD
76884-156 76884-158 76884-160
2,4-Difluoro-1-isopropoxybenzene ≥95%
2,4-Difluoro-1-isopropoxybenzene
Formula: C₉H₁₀F₂O
MW: 172.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070792
CAS Number: 203059-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR