4-(1-Piperidinyl)benzeneboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 1-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-piperidine

18949-1G 18949-25G 18949-5G
76884-146EA 66.84 USD
76884-146 76884-148 76884-150
4-(1-Piperidinyl)benzeneboronic acid pinacol ester ≥97% research grade
4-(1-Piperidinyl)benzeneboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₂
MW: 287.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368524
CAS Number: 852227-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR