2,4-Dichloro-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

18696-1G 18696-25G 18696-500MG 18696-5G
76884-106EA 292.81 USD
76884-106 76884-108 76884-110 76884-112
2,4-Dichloro-5-methylbenzoic acid ≥95%
2,4-Dichloro-5-methylbenzoic acid
Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03265315
CAS Number: 6660-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR