2-(4-Fluoro-2-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

18631-1G 18631-25G 18631-5G
76884-090EA 62.6 USD
76884-090 76884-092 76884-094
2-(4-Fluoro-2-methylphenyl)-1,3-dioxolane ≥97%
2-(4-Fluoro-2-methylphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06209321
CAS Number: 773093-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR