3-Bromo-2-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-iodo-3-methoxybenzene, 3-Bromo-2-iodophenyl methyl ether

18619-25G 18619-5G
76884-078EA 389.36 USD
76884-078 76884-080
3-Bromo-2-iodoanisole ≥97%
3-Bromo-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089327
CAS Number: 450412-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR