2,3-Difluoro-6-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18593-1G 18593-25G 18593-500MG 18593-5G
76884-064EA 379.81 USD
76884-064 76884-066 76884-068 76884-070
2,3-Difluoro-6-isopropoxyphenylboronic acid ≥97%
2,3-Difluoro-6-isopropoxyphenylboronic acid
Formula: C₉H₁₁BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28806014
CAS Number: 2121511-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR