1-Bromo-5-fluoro-4-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

18485-1G 18485-25G 18485-500MG 18485-5G
76884-034EA 192.03 USD
76884-034 76884-036 76884-038 76884-040
1-Bromo-5-fluoro-4-methoxy-2-methylbenzene ≥97%
1-Bromo-5-fluoro-4-methoxy-2-methylbenzene
Formula: C₈H₈BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD16658746
CAS Number: 550400-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR