2-(Benzyloxy)-1-bromo-3-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

18446-1G 18446-25G 18446-500MG 18446-5G
76884-022EA 253.56 USD
76884-022 76884-024 76884-026 76884-028
2-(Benzyloxy)-1-bromo-3-fluoro-4-methylbenzene ≥97%
2-(Benzyloxy)-1-bromo-3-fluoro-4-methylbenzene
Formula: C₁₄H₁₂BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD32631776
CAS Number: 2379321-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR