2-Benzyloxy-3,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)-3,5-difluorophenylboronic acid

18406-1G 18406-25G 18406-500MG 18406-5G
76884-006EA 130.5 USD
76884-006 76884-008 76884-010 76884-012
2-Benzyloxy-3,5-difluorophenylboronic acid ≥97%
2-Benzyloxy-3,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11855849
CAS Number: 1150114-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR