2,4-Dichloro-3-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

18383-1G 18383-25G 18383-500MG 18383-5G
76883-992EA 260.99 USD
76883-992 76883-994 76883-996 76883-998
2,4-Dichloro-3-fluorophenylboronic acid ≥95%
2,4-Dichloro-3-fluorophenylboronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23380952
CAS Number: 1160561-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR