2-(tert-Butylcarbonylamino)-6-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18338-1G 18338-25G 18338-500MG 18338-5G
76883-978EA 202.64 USD
76883-978 76883-980 76883-982 76883-984
2-(tert-Butylcarbonylamino)-6-chlorophenylboronic acid ≥97%
2-(tert-Butylcarbonylamino)-6-chlorophenylboronic acid
Formula: C₁₁H₁₅BClNO₃
MW: 255.506 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01114877
CAS Number: 1451390-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR