1-Bromo-3,5-difluoro-2-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

18312-1G 18312-25G 18312-500MG 18312-5G
76883-964EA 350.1 USD
76883-964 76883-966 76883-968 76883-970
1-Bromo-3,5-difluoro-2-(methoxymethoxy)benzene ≥97%
1-Bromo-3,5-difluoro-2-(methoxymethoxy)benzene
Formula: C₈H₇BrF₂O₂
MW: 253.041 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28246518
CAS Number: 1803589-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR