1-Isobutyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

18116-1G 18116-25G 18116-5G
76883-910EA 76.39 USD
76883-910 76883-912 76883-914
1-Isobutyl-1H-pyrazole ≥97%
1-Isobutyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.184 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08558977
CAS Number: 725746-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR