2,3-Difluoro-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18088-25G 18088-5G
76883-896EA 247.19 USD
76883-896 76883-898
2,3-Difluoro-4-methylphenylboronic acid ≥97%
2,3-Difluoro-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09751348
CAS Number: 508235-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR