1H-Indazole-7-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (1H-Indazol-7-yl)boronic acid

17857-1G 17857-25G 17857-5G 17857-500MG
76883-840EA 114.58 USD
76883-840 76883-842 76883-844 77649-012
1H-Indazole-7-boronic acid ≥97%
1H-Indazole-7-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09997801
CAS Number: 915411-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR