1-Fluoro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

17820-25G 17820-5G
76883-826EA 240.83 USD
76883-826 76883-828
1-Fluoro-2-isopropoxybenzene ≥97%
1-Fluoro-2-isopropoxybenzene
Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD18447593
CAS Number: 1160293-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR