1-Bromo-2,5-dichloro-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

17598-1G 17598-25G 17598-500MG 17598-5G
76883-800EA 171.87 USD
76883-800 76883-802 76883-804 76883-806
1-Bromo-2,5-dichloro-3-methoxybenzene ≥95%
1-Bromo-2,5-dichloro-3-methoxybenzene
Formula: C₇H₅BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD20483488
CAS Number: 174913-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR