2-(Methoxymethoxy)-3-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

17595-1G 17595-25G 17595-500MG 17595-5G
76883-788EA 225.98 USD
76883-788 76883-790 76883-792 76883-794
2-(Methoxymethoxy)-3-methylphenylboronic acid ≥97% research grade
2-(Methoxymethoxy)-3-methylphenylboronic acid
Formula: C₉H₁₃BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD23379601
CAS Number: 1104195-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR