2-(Propan-2-yloxy)naphthalene-1-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

17527-1G 17527-25G 17527-500MG 17527-5G
76883-760EA 194.15 USD
76883-760 76883-762 76883-764 76883-766
2-(Propan-2-yloxy)naphthalene-1-boronic acid ≥97% research grade
2-(Propan-2-yloxy)naphthalene-1-boronic acid
Formula: C₁₃H₁₅BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD12862849
CAS Number: 1084904-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR