N-(2,3-Dimethylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dimethylacetanilide

17488-1G 17488-25G 17488-500MG 17488-5G
76883-748EA 155.96 USD
76883-748 76883-750 76883-752 76883-754
N-(2,3-Dimethylphenyl)acetamide ≥97%
N-(2,3-Dimethylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
MW: 163.216 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00130064
CAS Number: 134-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR