2,4-Dibromo-6-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17447-1G 17447-25G 17447-5G
76883-722EA 62.6 USD
76883-722 76883-724 76883-726
2,4-Dibromo-6-fluorobenzoic acid ≥97%
2,4-Dibromo-6-fluorobenzoic acid
Formula: C₇H₃Br₂FO₂
MW: 297.90 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094087
CAS Number: 183065-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR