3-(Ethoxycarbonyl)-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid, 3-Fluoro-5-(ethoxycarbonyl)benzeneboronic acid

17324-1G 17324-25G 17324-5G 17324-500MG
76883-696EA 84.88 USD
76883-696 76883-698 76883-700 77648-974
3-(Ethoxycarbonyl)-5-fluorophenylboronic acid ≥97%
3-(Ethoxycarbonyl)-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MW: 211.9800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363792
CAS Number: 871329-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR