1-(Benzyloxy)-5-bromo-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

17314-25G 17314-5G
76883-688EA 350.1 USD
76883-688 76883-690
1-(Benzyloxy)-5-bromo-3-fluorobenzene ≥97%
1-(Benzyloxy)-5-bromo-3-fluorobenzene
Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181618
CAS Number: 130722-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR