2,4,6-Tribromophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17275-1G 17275-25G 17275-5G 17275-500MG
76883-674EA 109.28 USD
76883-674 76883-676 76883-678 77648-970
2,4,6-Tribromophenylboronic acid ≥97%
2,4,6-Tribromophenylboronic acid
Formula: C₆H₄BBr₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD13181641
CAS Number: 1451392-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR