1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(1,5-Dimethyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,5-Dimethyl-1H-pyrazole-4-boronic acid Pinacol ester

17199-25G 17199-5G
76883-630EA 456.19 USD
76883-630 76883-632
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥97%
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09864189
CAS Number: 1036991-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR