1-Bromo-3,5-Difluoro-2-Methylsulfanylbenzene ≥95%

Supplier: AOBChem USA

Warning

17178-1G 17178-25G 17178-500MG 17178-5G
76883-610EA 260.99 USD
76883-610 76883-612 76883-614 76883-616
1-Bromo-3,5-Difluoro-2-Methylsulfanylbenzene ≥95%
1-Bromo-3,5-Difluoro-2-Methylsulfanylbenzene
Formula: C₇H₅BrF₂S
Storage Temperature: Refrigerator
MDL Number: MFCD26045238
CAS Number: 861931-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR