1-Bromo-4-(methoxymethoxy)naphthalene ≥97% research grade

Supplier: AOBChem USA

Warning

17084-1G 17084-25G 17084-500MG 17084-5G
76883-596EA 225.98 USD
76883-596 76883-598 76883-600 76883-602
1-Bromo-4-(methoxymethoxy)naphthalene ≥97% research grade
1-Bromo-4-(methoxymethoxy)naphthalene
Formula: C₁₂H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31619385
CAS Number: 70654-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR