1,4-Dibromo-2-(trimethylsilyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

17076-25G 17076-5G
76883-584EA 310.85 USD
76883-584 76883-586
1,4-Dibromo-2-(trimethylsilyl)benzene ≥97% research grade
1,4-Dibromo-2-(trimethylsilyl)benzene
Formula: C₉H₁₂Br₂Si
MW: 308.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178301
CAS Number: 1032231-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR