1,5-Dibromo-3-fluoro-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

17002-25G 17002-5G
76883-518EA 524.09 USD
76883-518 76883-520
1,5-Dibromo-3-fluoro-2-(methoxymethoxy)benzene ≥95%
1,5-Dibromo-3-fluoro-2-(methoxymethoxy)benzene
Formula: C₈H₇Br₂FO₂
MW: 313.946 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32631772
CAS Number: 2379322-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR