1,3-Dibromo-5-fluoro-2-(phenylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

16992-25G 16992-5G
76883-510EA 364.95 USD
76883-510 76883-512
1,3-Dibromo-5-fluoro-2-(phenylmethoxy)benzene ≥97%
1,3-Dibromo-5-fluoro-2-(phenylmethoxy)benzene
Formula: C₁₃H₉Br₂FO
MW: 360.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00443143
CAS Number: 130046-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR