4-Bromo-2,3-dichloropyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,3,-Dichloro-4-bromopyridine

16917-1G 16917-25G 16917-500MG 16917-5G
76883-496EA 332.07 USD
76883-496 76883-498 76883-500 76883-502
4-Bromo-2,3-dichloropyridine ≥95%
4-Bromo-2,3-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MW: 226.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743493
CAS Number: 1020717-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR