2-(4-Bromo-2-methoxyphenyl)-[1,3]dioxolane ≥97%

Supplier: AOBChem USA

Warning

16880-1G 16880-25G 16880-5G
76883-486EA 64.72 USD
76883-486 76883-488 76883-490
2-(4-Bromo-2-methoxyphenyl)-[1,3]dioxolane ≥97%
2-(4-Bromo-2-methoxyphenyl)-[1,3]dioxolane
Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28368830
CAS Number: 109142-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR