5'-Bromo-2,2,2,2'-tetrafluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanone, 1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethan-1-one

16747-1G 16747-25G 16747-500MG 16747-5G
76883-466EA 192.03 USD
76883-466 76883-468 76883-470 76883-472
5'-Bromo-2,2,2,2'-tetrafluoroacetophenone ≥95%
5'-Bromo-2,2,2,2'-tetrafluoroacetophenone
Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547807
CAS Number: 617706-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR