1,2-Difluoro-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

16652-1G 16652-25G 16652-500MG 16652-5G
76883-442EA 124.13 USD
76883-442 76883-444 76883-446 76883-448
1,2-Difluoro-4-isopropoxybenzene ≥97%
1,2-Difluoro-4-isopropoxybenzene
Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070791
CAS Number: 203059-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR