2,3-Dimethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

16619-25G 16619-5G
76883-430EA 190.97 USD
76883-430 76883-432
2,3-Dimethoxyphenol ≥97%
2,3-Dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00008366
CAS Number: 5150-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR