2,3-Dichloro-6-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dichloro-6-trifluoromethylbenzaldehyde

16481-25G 16481-5G
76883-404EA 310.85 USD
76883-404 76883-406
2,3-Dichloro-6-(trifluoromethyl)benzaldehyde ≥95%
2,3-Dichloro-6-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD06660273
CAS Number: 186517-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR