1,3-Bibromo-5-propoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

16462-1G 16462-25G 16462-500MG 16462-5G
76883-388EA 264.17 USD
76883-388 76883-390 76883-392 76883-394
1,3-Bibromo-5-propoxybenzene ≥97% research grade
1,3-Bibromo-5-propoxybenzene
Formula: C₉H₁₀Br₂O
MW: 293.983 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28976873
CAS Number: 1579943-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR