(2,3-Difluoro-4-hydroxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

16419-1G 16419-25G 16419-500MG 16419-5G
76883-374EA 165.51 USD
76883-374 76883-376 76883-378 76883-380
(2,3-Difluoro-4-hydroxyphenyl)boronic acid ≥97%
(2,3-Difluoro-4-hydroxyphenyl)boronic acid
Formula: C₆H₅BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09996974
CAS Number: 1261169-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR