(3-Bromo-5-fluorophenoxy)(tert-butyl)dimethylsilane ≥97%

Supplier: AOBChem USA

Warning

16381-1G 16381-25G 16381-500MG 16381-5G
76883-360EA 203.7 USD
76883-360 76883-362 76883-364 76883-366
(3-Bromo-5-fluorophenoxy)(tert-butyl)dimethylsilane ≥97%
(3-Bromo-5-fluorophenoxy)(tert-butyl)dimethylsilane
Formula: C₁₂H₁₈BrFOSi
MW: 305.259 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29086253
CAS Number: 1310697-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR