2-(2,3-Difluorophenyl)[1,3]dioxolane ≥97%

Supplier: AOBChem USA

Warning

16352-25G 16352-5G
76883-350EA 466.8 USD
76883-350 76883-352
2-(2,3-Difluorophenyl)[1,3]dioxolane ≥97%
2-(2,3-Difluorophenyl)[1,3]dioxolane
Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209062
CAS Number: 773101-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR