2,3-Difluoro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16350-1G 16350-25G 16350-500MG 16350-5G
76883-334EA 171.87 USD
76883-334 76883-336 76883-338 76883-340
2,3-Difluoro-4-iodobenzaldehyde ≥97%
2,3-Difluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₂IO
MW: 268.0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09835190
CAS Number: 885590-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR