2,4-Dibromo-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16296-1G 16296-25G 16296-5G
76883-324EA 92.3 USD
76883-324 76883-326 76883-328
2,4-Dibromo-6-fluorobenzaldehyde ≥97%
2,4-Dibromo-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Br₂FO
MW: 281.905 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094089
CAS Number: 205683-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR