1,2-Dimethoxy-4-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

16098-25G 16098-5G
76883-316EA 346.92 USD
76883-316 76883-318
1,2-Dimethoxy-4-(methoxymethoxy)benzene ≥95%
1,2-Dimethoxy-4-(methoxymethoxy)benzene
Formula: C₁₀H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD27943688
CAS Number: 133129-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR