1-Bromo-3,4-difluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

16032-1G 16032-25G 16032-5G
76883-306EA 94.43 USD
76883-306 76883-308 76883-310
1-Bromo-3,4-difluoro-2-methoxybenzene ≥97%
1-Bromo-3,4-difluoro-2-methoxybenzene
Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295239
CAS Number: 888318-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR