2,4-Dichloro-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16001-1G 16001-25G 16001-500MG 16001-5G
76883-290EA 165.51 USD
76883-290 76883-292 76883-294 76883-296
2,4-Dichloro-3-methoxyphenylboronic acid ≥97%
2,4-Dichloro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD10699638
CAS Number: 944128-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR