(6-Ethoxy-2,3-difluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

15988-1G 15988-25G 15988-5G
76883-278EA 64.72 USD
76883-278 76883-280 76883-282
(6-Ethoxy-2,3-difluorophenyl)boronic acid ≥97%
(6-Ethoxy-2,3-difluorophenyl)boronic acid
Formula: C₈H₉BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD12026698
CAS Number: 1309980-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR